

1-(2-Bromoethynyl)-4-chloro-2-fluorobenzene
Catalog # APOSPC400027-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:233.465 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:2187435-46-5
Formula:C₈H₃BrClF
MDL Number:MFCD31536745