
Ethyl 7-cyano-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1555847-50-1
:
Formula:C₁₂H₈F₃N₃O₂
MDL Number:MFCD32200426
:283.206 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together