4,6-Difluoro-4,6-dideoxy-D-galactopyranose 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:238403-53-7
:
Formula:C₆H₁₀F₂O₄
MDL Number:MFCD08461578
:184.138 g/mol
Melting Pt:169…177 °C
Storage Temperature:Ambient
Frequently Bought Together