
Ethyl 2-hydroxy-2-(trifluoromethyl)propanoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:107018-39-3
:
Formula:C₆H₉F₃O₃
MDL Number:MFCD09258952
Boiling Pt:85 °C (150 mm Hg)
:186.129 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together