
(1R)-1-(Trifluoromethyl)ethylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:177469-12-4
:
Formula:C₃H₆F₃N·ClH
MDL Number:MFCD07779095
:149.543 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together