
2',3'-Difluoro-4'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:237761-82-9
:
Formula:C₉H₅F₅O
MDL Number:MFCD00236252
:224.127 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together