
1-[2-Amino-4-(trifluoromethyl)phenyl]-3,3-dimethylazetidin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:861881-17-6
:
Formula:C₁₂H₁₃F₃N₂O
MDL Number:MFCD09907668
:258.24 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together