

2,3-Difluoro-4-hydroxybenzotrifluoride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:116640-12-1
:
Formula:C₇H₃F₅O
MDL Number:MFCD00236266
:198.09 g/mol
UN:1760
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together