
2-(Difluoromethyl)-5-nitro-1H-benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:97273-25-1
:
Formula:C₈H₅F₂N₃O₂
MDL Number:MFCD18909881
:213.141 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together