
4-Bromo-2-(difluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1388056-90-3
:
Formula:C₈H₅BrF₂N₂
MDL Number:MFCD22561277
:247.04 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together