
2-(Trifluoromethyl)-4-nitro-1H-benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14689-51-1
:
Formula:C₈H₄F₃N₃O₂
MDL Number:MFCD00796063
:231.131 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together