
4-(2-Fluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1065074-15-8
:
Formula:C₈H₆FN₃O
MDL Number:MFCD11504880
:179.151 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together