Order Entry
Czech Republic
ContactUsLinkComponent
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Irritant
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Catalog # APOSPC32883-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Catalog # APOSPC32883-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.567 g/mol
Melting Pt:191 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:478066-06-7
Formula:C₇H₃ClF₃N₃
MDL Number:MFCD01315052