
1-Bromo-4-(2,2-difluorocyclopropyl)benzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1275621-14-1
:
Formula:C₉H₇BrF₂
MDL Number:MFCD22548234
:233.053 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together