7-(Trifluoromethyl)chroman-4-one 97%
Catalog # APOSPC3277-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Flash Pt:>110 °C
:216.157 g/mol
Storage Temperature:Ambient
CAS Number:111141-02-7
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD09998165