1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-1H-pyrrole
Catalog # APOSPC32453-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.073 g/mol
Storage Temperature:Ambient
CAS Number:259175-24-1
Formula:C₁₁H₆Cl₂F₃N
MDL Number:MFCD00278514