2-(Hydroxymethyl)-4-phenyl-5-(trifluoromethyl)thiophene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:256488-46-7
:
Formula:C₁₂H₉F₃OS
MDL Number:MFCD01934530
:258.259 g/mol
Storage Temperature:Ambient
Frequently Bought Together