
(1S)-1-(3-Fluorophenyl)ethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:444643-09-8
:
Formula:C₈H₁₀FN
MDL Number:MFCD06761860
:139.17 g/mol
Boiling Pt:63…67 °C (4.2 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together