Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-N-(5,7-difluoro-1,3-benzothiazol-2-yl)acetamide
Irritant
2-Bromo-N-(5,7-difluoro-1,3-benzothiazol-2-yl)acetamide
Catalog # APOSPC31221-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Bromo-N-(5,7-difluoro-1,3-benzothiazol-2-yl)acetamide
Catalog # APOSPC31221-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :307.115 g/mol
Melting Pt:191…193 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:219499-50-0
Formula:C₉H₅BrF₂N₂OS
MDL Number:MFCD00177782