Order Entry
Czech Republic
ContactUsLinkComponent
N1-[5-(Trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
N1-[5-(Trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Catalog # APOSPC31127-1G
Supplier:  Apollo Scientific
CAS Number:  
N1-[5-(Trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Catalog # APOSPC31127-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.18 g/mol
Storage Temperature:Ambient
CAS Number:207557-34-4
Formula:C₈H₁₀F₃N₃
MDL Number:MFCD00816592