
Ethyl 8-fluoro-2-(trifluoromethyl)imidazo(1,2-a)pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2379918-41-7
:
Formula:C₁₁H₈F₄N₂O₂
MDL Number:MFCD32066750
:276.187 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together