
3,6-Difluoro-2-(trifluoromethoxy)aniline 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1806353-36-5
:
Formula:C₇H₄F₅NO
MDL Number:MFCD28790983
:213.105 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together