3-(Difluoromethoxy)phenacyl bromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:443914-96-3
:
Formula:C₉H₇BrF₂O₂
MDL Number:MFCD09259003
ADR:8,III
:265.051 g/mol
UN:1759
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together