3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2324179-05-5
:
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD32640178
:283.042 g/mol
Storage Temperature:Ambient
Frequently Bought Together