3-Bromo-2-chloro-5-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303598-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:357.96 g/mol
Storage Temperature:Ambient
CAS Number:2384426-01-9
Formula:C₇H₂BrCl₂F₃O₂S
MDL Number:MFCD32640171