Order Entry
Czech Republic
ContactUsLinkComponent
4-Phenoxy-2-(trifluoromethyl)benzoic acid 97%
4-Phenoxy-2-(trifluoromethyl)benzoic acid 97%
Catalog # APOSPC303551-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Phenoxy-2-(trifluoromethyl)benzoic acid 97%
Catalog # APOSPC303551-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :282.215 g/mol
Storage Temperature:Ambient
CAS Number:1253188-35-0
Formula:C₁₄H₉F₃O₃
MDL Number:MFCD32189207