5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1881295-62-0
:
Formula:C₈H₃BrF₄O₂
MDL Number:MFCD29043332
:287.006 g/mol
Storage Temperature:Ambient
Frequently Bought Together