3-Bromo-6-fluoro-2-(trifluoromethyl)phenylacetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2385598-79-6
:
Formula:C₉H₅BrF₄O₂
MDL Number:MFCD32640120
:301.032 g/mol
Storage Temperature:Ambient
Frequently Bought Together