2-Bromo-4,6-bis(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1807224-31-2
:
Formula:C₉H₄Br₂F₆
MDL Number:MFCD28728005
:385.926 g/mol
Storage Temperature:Ambient
Frequently Bought Together