Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-4,6-bis(trifluoromethyl)benzaldehyde 97%
2-Bromo-4,6-bis(trifluoromethyl)benzaldehyde 97%
Catalog # APOSPC303528-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-4,6-bis(trifluoromethyl)benzaldehyde 97%
Catalog # APOSPC303528-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :321.014 g/mol
Storage Temperature:Ambient
CAS Number:1415130-39-0
Formula:C₉H₃BrF₆O
MDL Number:MFCD28727819