Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzaldehyde 97%
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzaldehyde 97%
Catalog # APOSPC303498-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzaldehyde 97%
Catalog # APOSPC303498-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :321.906 g/mol
Storage Temperature:Ambient
Formula:C₈H₂BrCl₂F₃O
MDL Number:MFCD32640091