3-Bromo-2-fluoro-5-(trifluoromethyl)benzamide 97%
Catalog # APOSPC303472-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.021 g/mol
Storage Temperature:Ambient
CAS Number:2092800-03-6
Formula:C₈H₄BrF₄NO
MDL Number:MFCD32265222