3-Bromo-2-fluoro-5-(trifluoromethyl)benzoyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2168326-54-1
:
Formula:C₈H₂BrClF₄O
MDL Number:MFCD32640079
:305.451 g/mol
Storage Temperature:Ambient
Frequently Bought Together