3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2091606-48-1
:
Formula:C₈H₃BrF₄O₂
MDL Number:MFCD31382551
:287.006 g/mol
Storage Temperature:Ambient
Frequently Bought Together