Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxy-4-(trifluoromethoxy)cinnamic acid 97%
2-Methoxy-4-(trifluoromethoxy)cinnamic acid 97%
Catalog # APOSPC303402-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-4-(trifluoromethoxy)cinnamic acid 97%
Catalog # APOSPC303402-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.182 g/mol
Storage Temperature:Ambient
CAS Number:1262019-21-5
Formula:C₁₁H₉F₃O₄
MDL Number:MFCD18398498