2-Chloro-4-fluoro-5-methylbenzoyl chloride
Catalog # APOSPC303343-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.029 g/mol
Storage Temperature:Ambient
CAS Number:1807027-18-4
Formula:C₈H₅Cl₂FO
MDL Number:MFCD28751941