Order Entry
Czech Republic
ContactUsLinkComponent
 
2-Chloro-4-fluoro-5-methylphenylacetonitrile 97%
Catalog # APOSPC303340-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-fluoro-5-methylphenylacetonitrile 97%
Catalog # APOSPC303340-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.61 g/mol
Storage Temperature:Ambient
CAS Number:1000513-29-0
Formula:C₉H₇ClFN
MDL Number:MFCD09923912