4-Chloro-2-fluoro-5-methylbenzenesulphonyl chloride
Catalog # APOSPC303329-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.083 g/mol
Storage Temperature:Ambient
CAS Number:868755-70-8
Formula:C₇H₅Cl₂FO₂S
MDL Number:MFCD08060548