4-Chloro-2-fluoro-5-methylbenzoyl chloride
Catalog # APOSPC303326-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.029 g/mol
Storage Temperature:Ambient
CAS Number:204778-68-7
Formula:C₈H₅Cl₂FO
MDL Number:MFCD13174586