6-Chloro-2,4-difluoro-3-methylphenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1858250-05-1
:
Formula:C₉H₆ClF₂N
MDL Number:MFCD29038430
:201.6 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together