4-Butoxy-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC303271-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.707 g/mol
Storage Temperature:Ambient
CAS Number:1706431-01-7
Formula:C₁₀H₁₁ClF₂O₃S
MDL Number:MFCD28054242