Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-2-methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
3-Bromo-2-methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
Catalog # APOSPC303265-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-2-methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
Catalog # APOSPC303265-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :395.592 g/mol
Storage Temperature:Ambient
CAS Number:1706431-07-3
Formula:C₇H₅BrClF₅O₂S₂
MDL Number:MFCD28054247