
2-Methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
Catalog # APOSPC303263-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:316.696 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:1706458-82-3
Formula:C₇H₆ClF₅O₂S₂
MDL Number:MFCD28054235