4-Fluoro-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303261-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.636 g/mol
Storage Temperature:Ambient
CAS Number:1706458-75-4
Formula:C₈H₅ClF₄O₂S
MDL Number:MFCD28054246