3-Chloro-5-fluoro-4-methoxybenzenesulphonyl chloride
Catalog # APOSPC303249-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.082 g/mol
Storage Temperature:Ambient
CAS Number:1706458-23-2
Formula:C₇H₅Cl₂FO₃S
MDL Number:MFCD28054214