3,4-Dichloro-5-(trifluoromethoxy)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706458-58-3
:
Formula:C₉H₄Cl₂F₃NO
MDL Number:MFCD28054208
:270.035 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together