6-Chloro-2,3-difluorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1208078-24-3
:
Formula:C₆H₂Cl₂F₂O₂S
MDL Number:MFCD12922581
:247.047 g/mol
Storage Temperature:Ambient
Frequently Bought Together