2,4-Bis(trifluoromethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:376637-88-6
:
Formula:C₈H₃ClF₆O₂S
MDL Number:MFCD13185342
:312.617 g/mol
Storage Temperature:Ambient
Frequently Bought Together