5-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:915763-87-0
:
Formula:C₇H₃ClF₄O₂S
MDL Number:MFCD13185646
:262.609 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
Frequently Bought Together