3,4-Dichloro-5-(trifluoromethoxy)benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706458-14-1
:
Formula:C₈H₂Cl₂F₃NO
MDL Number:MFCD28054210
:256.009 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together